EMAIL THIS PAGE TO A FRIEND
PS2025 Supelco

Phenthoate

analytical standard

  • CAS Number 2597-03-7

  • Empirical Formula (Hill Notation) C12H17O4PS2

  • Molecular Weight 320.36

  •  Beilstein Registry Number 2474108

  •  EC Number 219-997-0

  •  MDL number MFCD00145099

  •  PubChem Substance ID 24899136

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Insecticides, Organophorous, Pesticides & Metabolites Standards More...
grade   analytical standard
InChI Key   XAMUDJHXFNRLCY-UHFFFAOYSA-N
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-2025
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN2810 - class 6.1 - PG 3 - SMP - EHS - Toxic, liquids, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
AI7875000

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of 'Positive List' Chemicals (Mix 1) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of 'Positive List' Chemicals (Mix 1) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Herbicides, Pesticides, Purification

GC Analysis of Organophosphorus Pesticides (OP-Pest) in Cabbage, Onion, and Mushrooms on SLB-5ms after SPE using Supelclean™ ENVI-Carb™ II/PSA

From our library of Articles, Sigma-Aldrich presents GC Analysis of Organophosphorus Pesticides (OP-Pest) in Cabbage, Onion, and Mushrooms on SLB-5ms after SPE using Supelclean™ ENVI-Carb™ II/PSA
Keywords: Agriculture, Centrifugation, Chromatography, Degradations, Environmental, Gas chromatography, Nitrogen phosphorus detector, Pesticides, Purification, Sample preparations, Separation, Solid phase extractions

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?