EMAIL THIS PAGE TO A FRIEND
PS2041 Supelco

Flusilazole

analytical standard

  • CAS Number 85509-19-9

  • Empirical Formula (Hill Notation) C16H15F2N3Si

  • Molecular Weight 315.39

  •  Beilstein Registry Number 5824097

  •  MDL number MFCD01076615

  •  PubChem Substance ID 24899142

Purchase

Properties

Related Categories All Fungicides, Analytical Standards, Analytical/Chromatography, Conazoles, Food and Cosmetic Component Standards,
grade   analytical standard
InChI Key   FQKUGOMFVDPBIZ-UHFFFAOYSA-N
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-2041
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
XZ4105000

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of 'Positive List' Chemicals (Mix 4) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of 'Positive List' Chemicals (Mix 4) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Herbicides, Pesticides, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?