EMAIL THIS PAGE TO A FRIEND
PS2043 Supelco

Imazalil

analytical standard

Synonym: 1-[2-(Allyloxy)-2-(2,4-dichlorophenyl)ethyl]imidazole

  • CAS Number 35554-44-0

  • Empirical Formula (Hill Notation) C14H14Cl2N2O

  • Molecular Weight 297.18

  •  Beilstein Registry Number 545683

  •  EC Number 252-615-0

  •  MDL number MFCD00055331

  •  PubChem Substance ID 329823645

Purchase

Properties

Related Categories All Fungicides, Analytical Standards, Analytical/Chromatography, Conazoles, Fungicides,
grade   analytical standard
InChI Key   PZBPKYOVPCNPJY-UHFFFAOYSA-N
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-2043
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2811 6.1/PG 3
WGK Germany 
3
RTECS 
NI4776000

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of Agricultural Pesticides (Standard) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of Agricultural Pesticides (Standard) on SLB®-5ms
Keywords: Agriculture, Chromatography, Gas chromatography, Gas chromatography mass spectrometry, Herbicides, Mass spectrometry, Pesticides

GC Analysis of Agriculture Pesticides in Orange (Spiked) on SLB®-5ms after QuEChERS Cleanup using Primary-Secondary Amine (PSA)

From our library of Articles, Sigma-Aldrich presents GC Analysis of Agriculture Pesticides in Orange (Spiked) on SLB®-5ms after QuEChERS Cleanup using Primary-Secondary Amine (PSA)
Keywords: Chromatography, Gas chromatography, Pesticides, Purification, Sample preparations, Solid phase extractions

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?