EMAIL THIS PAGE TO A FRIEND
PS702 Supelco

Mirex

analytical standard

Synonym: Perchloropentacyclodecane

  • CAS Number 2385-85-5

  • Empirical Formula (Hill Notation) C10Cl12

  • Molecular Weight 545.54

  •  Beilstein Registry Number 5605269

  •  EC Number 219-196-6

  •  MDL number MFCD00167995

  •  PubChem Substance ID 24899280

Purchase

Properties

grade   analytical standard
InChI Key   GVYLCNUFSHDAAW-UHFFFAOYSA-N
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-702
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN2811 - class 6.1 - PG 3 - MP - EHS - Toxic solids, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
PC8225000

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Carcinogen Selector - Digestive System - Liver

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Digestive System - Liver
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System - Adrenal Gland

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Endocrine System - Adrenal Gland
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System - Liver Endocrine

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Endocrine System - Liver Endocrine
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Endocrine System
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Hematopoietic System

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Hematopoietic System
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Urinary System - Kidney

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Urinary System - Kidney
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Urinary System

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Urinary System
Keywords: Cancer, Carcinogens, Respiratory

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?