-
Product Directory
Custom Product
-
Services Offered
Custom Capabilities
-
Synonym:
1,4,5,6,7,8,8-
CAS Number 76-44-8
Empirical Formula (Hill Notation) C10H5Cl7
Molecular Weight 373.32
EC Number 200-962-3
| Related Categories | 2000/60/EC, Alphabetic, Analytical Standards, Analytical/Chromatography, Chromatography, |
| grade | analytical standard |
| packaging | ampule of 100 mg |
| mfr. no. | Chem Service Inc, PS-78 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?