EMAIL THIS PAGE TO A FRIEND
SBSI13 Supelco

SULFUR IN ISOOCTANE

analytical standard, 0-0.300 wt. % in isooctane

Purchase

Properties

Related Categories ASTM D2622, ASTM D6334, ASTM Petroleum Standards, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
packaging   set of 13 × 100 mL
concentration   0-0.300 wt. % in isooctane
application(s)   ICP/AAS: suitable
suitability   suitable for D2622 per ASTM
  suitable for D6334 per ASTM
format   matrix material
storage temp.   room temp

Description

Analysis Note

Concentration Table
Sulfur, Wt.%
• Standard 1 0.0000
• Standard 2 0.0010
• Standard 3 0.0020
• Standard 4 0.0030
• Standard 5 0.0050
• Standard 6 0.0100
• Standard 7 0.0200
• Standard 8 0.0300
• Standard 9 0.0400
• Standard 10 0.0600
• Standard 11 0.1000
• Standard 12 0.2000
• Standard 13 0.3000

Application

For gasoline and reformulated gasoline analysis using X-ray fluorescence.

General description

Di-n-butyl sulfide in Isooctane.

Price and Availability


Nylon Membrane and Net Filters
Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1262 - class 3 - PG 2 - Octanes
WGK Germany 
2
Flash Point(F) 
10.4 °F
Flash Point(C) 
-12 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?