• USA Home
  • SDF7 - SULFUR IN DIESEL FUEL SET, 0-0.1WT%

EMAIL THIS PAGE TO A FRIEND
SDF7 Supelco

SULFUR IN DIESEL FUEL SET, 0-0.1WT%

analytical standard, set of 7 × 100 mL

Synonym: Sulfur in Synthetic Diesel Fuel

Purchase

Properties

Related Categories ASTM D4294, ASTM Petroleum Standards, Analytical Standards, Analytical/Chromatography, Lubricating Oils,
grade   analytical standard
packaging   set of 7 × 100 mL
concentration   0-0.100 wt. %
application(s)   ICP/AAS: suitable
suitability   suitable for D2622 per ASTM
  suitable for D4294 per ASTM
  suitable for D7039 per ASTM
format   matrix material
storage temp.   room temp

Description

Analysis Note

Concentration Table
Sulfur, Wt.%
• Standard 1 0.0000
• Standard 2 0.0200
• Standard 3 0.0400
• Standard 4 0.0500
• Standard 5 0.0600
• Standard 6 0.0800
• Standard 7 0.1000

General description

Di-N-butyl sulfide in synthetic diesel fuel.

Price and Availability


Nylon Membrane and Net Filters
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
RIDADR 
UN1202 - class 3 - PG 3 - Diesel fuel
WGK Germany 
2
Flash Point(F) 
>120.2 °F
Flash Point(C) 
>49 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?