• USA Home
  • SIO11 - SULFUR IN ISOOCTANE SET, 0-1000 NG/UL

EMAIL THIS PAGE TO A FRIEND
SIO11 Supelco

SULFUR IN ISOOCTANE SET, 0-1000 NG/UL

analytical standard, set of 11 × 100 mL

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Lubricating Oils, Petrochemical Standards, Sulfur Standards More...
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
packaging   set of 11 × 100 mL
concentration   0-1000.00 ng/μL in isooctane
application(s)   ICP/AAS: suitable
suitability   suitable for D5453 per ASTM
format   matrix material
storage temp.   room temp

Description

Analysis Note

Concentration Table
Sulfur, Wt.%
• Standard 1 0.0000
• Standard 2 0.5000
• Standard 3 1.0000
• Standard 4 2.5000
• Standard 5 5.0000
• Standard 6 10.000
• Standard 7 25.000
• Standard 8 50.000
• Standard 9 100.00
• Standard 10 500.00
• Standard 11 1000.0

General description

Di-N-butyl sulfide in Isooctane.

Price and Availability


Nylon Membrane and Net Filters
Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
RIDADR 
UN1262 - class 3 - PG 2 - Octanes, solution
WGK Germany 
2
Flash Point(F) 
10.4 °F
Flash Point(C) 
-12 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?