• USA Home
  • SNVRO12 - Sulfur and metals in residual fuel

EMAIL THIS PAGE TO A FRIEND
SNVRO12 Supelco

Sulfur and metals in residual fuel

analytical standard, set of 12 × 100 mL

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Petrochemical Standards, Sulfur Standards
grade   analytical standard
packaging   set of 12 × 100 mL
application(s)   ICP/AAS: suitable
format   matrix material
storage temp.   room temp

Description

Analysis Note

Concentration Table
. . . . . . . .Sulfur, Wt.% . . . Ni (ppm) . . . . . V (ppm)
• Standard 1 0.1850 . . . . . . 4 . . . . . . . . . . .1
• Standard 2 0.5000 . . . . . . 10 . . . . . . . . . .500
• Standard 3 1.0000 . . . . . . 100 . . . . . . . . .25
• Standard 4 1.5000 . . . . . . 80 . . . . . . . . . .250
• Standard 5 2.0000 . . . . . . 40 . . . . . . . . . .100
• Standard 6 2.5000 . . . . . . . 5 . . . . . . . . . . .400
• Standard 7 3.0000 . . . . . . . 60 . . . . . . . . . . 300
• Standard 8 3.5000 . . . . . . . 4 . . . . . . . . . . . 200
• Standard 9 4.0000 . . . . . . . 100 . . . . . . . . ..1
• Standard 10 4.5000 . . . . . . . 50 . . . . . . . . . . 250
• Standard 11 5.0000 . . . . . . . 20 . . . . . . . . . . 500
• Standard 12 5.5000 . . . . . . . 100 . . . . . . . . . 50

General description

Same as SNVMO except that the matrix is Residual Fuel Oil. Concentration of Ni and V are independent of the sulfur concentration. 12 standards per set: 100 mL each.

Price and Availability


Nylon Membrane and Net Filters

Analyte

Description

Product #

Add to Cart

Nickel 5-100 ppm SDS    
Vanadium 25-500 ppm SDS    
Sulfur .5-5.5 wt. % SDS    
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1202 - class 3 - PG 3 - Diesel fuel
WGK Germany 
3
Flash Point(F) 
249.8 °F
Flash Point(C) 
121 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?