• USA Home
  • SRO10 - SULFUR IN RESIDENTIAL OIL 0.25-5.0WT%

EMAIL THIS PAGE TO A FRIEND
SRO10 Supelco

SULFUR IN RESIDENTIAL OIL 0.25-5.0WT%

analytical standard, set of 10 × 100 mL

Purchase

Properties

Related Categories ASTM D2622, ASTM D4294, ASTM Petroleum Standards, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
packaging   set of 10 × 100 mL
concentration   0.25-5.00 wt. %
application(s)   ICP/AAS: suitable
suitability   suitable for D2622 per ASTM
  suitable for D4294 per ASTM
format   matrix material
storage temp.   room temp

Description

Analysis Note

Concentration Table
Sulfur, Wt.%
• Standard 1 0.2500
• Standard 2 0.5000
• Standard 3 1.0000
• Standard 4 1.5000
• Standard 5 2.0000
• Standard 6 2.5000
• Standard 7 3.0000
• Standard 8 3.5000
• Standard 9 4.0000
• Standard 10 5.0000

General description

Di-N-butyl sulfide in residual oil.

Price and Availability


Nylon Membrane and Net Filters
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1202 - class 3 - PG 3 - Diesel fuel
WGK Germany 
3
Flash Point(F) 
147.2 - 590 °F
Flash Point(C) 
64 - 310 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?