• USA Home
  • ULSD0-25 - ULTRA LOW SULFUR IN DIESEL OIL 0-25PPM

EMAIL THIS PAGE TO A FRIEND
ULSD0-25 Supelco

ULTRA LOW SULFUR IN DIESEL OIL 0-25PPM

set of 6 × 100 mL

Purchase

Properties

packaging   set of 6 × 100 mL
concentration   0-25 ppm
storage temp.   room temp

Description

General description

Sulfur with Ultra Low Diesel Fuel. Concentrations of 0, 5, 10, 15, 20 and 25 ppm. 100 mL each.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?