• USA Home
  • Z226025 - Spherisorb® Si HPLC Column
EMAIL THIS PAGE TO A FRIEND
Z226025 Supelco
Spherisorb® Si HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
  •  eCl@ss 32110501
Purchase
Properties
Related Categories Analytical/Chromatography, Established Column Lines from Other Manufacturers, HPLC, HPLC Columns, by Brand for Small Molecules, Si,
parameter   20-45 °C temp. range
L × I.D.   25 cm × 4.6 mm
matrix   silica
matrix active group   silica phase
particle size   5 μm
pore size   80 Å
surface area   220 m2/g
mode of chromatography   Hydrophilic interaction (HILIC)
  Normal phase
Description
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Spherisorb is a registered trademark of Waters
Price and Availability
All labs need water
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Content
Supelco HPLC Calculator for Isocratic or Gradient Method Transfer
Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography
Related Products
guard cartridge
Product #
Description
Add to Cart
Z226963 Spherisorb® Si Guard Cartridge, 5 μm particle size, L × I.D. 1 cm × 4.6 mm
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?