EMAIL THIS PAGE TO A FRIEND
Z226548 Supelco
Column Frits
diam. 1/4 in., pore size 2 μm
Purchase
Properties
Related Categories Analytical/Chromatography, Column Hardware, Column frits, 1/4 inch, HPLC, HPLC Accessories,
description   for Conventional Column Hardware (compression style)
packaging   pkg of 5 ea
diam.   1/4 in.
pore size   2 μm
Description
General description
Choose frits with 0.5 μm pores to retain packings prepared from 3 μm particles, frits with 2 μm pores to retain larger particles.
Price and Availability
All labs need water
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Content
Supelco HPLC Calculator for Isocratic or Gradient Method Transfer
Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?