• USA Home
  • Z227048 - Spherisorb® SAX Guard Cartridge
EMAIL THIS PAGE TO A FRIEND
Z227048 Supelco
Spherisorb® SAX Guard Cartridge
5 μm particle size, L × I.D. 1 cm × 4.6 mm
Purchase
Properties
packaging   pkg of 4 ea
L × I.D.   1 cm × 4.6 mm
matrix active group   ammonium, quaternary phase
particle size   5 μm
compatibility   for use with 3.0 to 4.6 mm column
Description
Legal Information
Spherisorb is a registered trademark of Waters
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
required but not provided
Product #
Description
Add to Cart
54987 Holder and Coupler for Legacy Guard Columns, for use with non-Supelco guard cartridges (not TSKgel guard cartridges)
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?