• USA Home
  • 1268885 - Etoricoxib Related Compound B

EMAIL THIS PAGE TO A FRIEND
1268885 USP

Etoricoxib Related Compound B

United States Pharmacopeia (USP) Reference Standard

Synonym: 4-(Methylsulfonyl)phenylacetic acid

Purchase

Properties

InChI Key   HGGWOSYNRVOQJH-UHFFFAOYSA-N
mp   136-140 °C(lit.)

Description

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Other Notes

Sales restrictions may apply.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?