Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
259853
Aldrich
Castor oil
CAS Number
8001-79-4
MDL number
MFCD00130746
EG/EC Number
232-293-8
Expand/Collapse All
Price and Availability
Descriptions
Packaging
1 L in glass btl
5, 250 mL in glass btl
Properties
optical activity
[α]20/D +5°, c = 5 in ethanol
refractive index
n
20/D
1.478(lit.)
bp
313 °C(lit.)
density
0.961 g/mL at 25 °C
References
Merck
Merck
13
,1908
reference
Corp MSDS
1
(1), 701:C /
FT-IR
2
(1), 998:A /
FT-IR
1
(1), 621:D /
Sax
6
, 654 /
Sigma FT-IR
1
(1), 1485:B
Safety
WGK Germany
-
RTECS
FI4100000
Flash Point(F)
235 °F
Flash Point(C)
113 °C
F
10
Related Categories
...
Essential Biological Reagents
>
Lipids and Related Products
...
Lipids
>
Oils
Scroll Up
Scroll Down
Related Information
FT-IR Raman
MSDS
Specification Sheet
Certificate of Analysis
Certificate of Origin
More Information
Links
Similar Products
Related Categories
Page Options
Printer Friendly View
Ask A Scientist
Email Page
Last 5 Products Viewed
259853 (Aldrich)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.