|
| 327123 Aldrich |
Liquid scintillation mixturePPO 100 g/L, POPOP 1.25 g/L, in toluene |
|
|||||||
| Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. |