General description
Shows the ability to generate chemically an electric current. A device is equipped with magnesium and copper electrodes, which are connected to a camera flash bulb. Immersion of flash device electrodes in hydrochloric acid solution generates a current, activating the flash. Eight demonstrations per kit.
Fundamental chemical principles are best retained by students when visual aids are utilized. The CHEMDEMO kits allow educators to present the basic properties of chemistry safely and easily.