General description
In this kit, different means of metallic iron oxidation are demonstrated by using an Agar gel. They include oxidation of iron using an external power supply such as a battery, spontaneous oxidation of iron, and inhibition of spontaneous oxidation of iron with a magnesium strip. One demonstration per kit.
Fundamental chemical principles are best retained by students when visual aids are utilized. The CHEMDEMO kits allow educators to present the basic properties of chemistry safely and easily.