Other Notes
Pure-Pac® II containers require the Micromatic MacroValve coupler for dispensing solvents, Z560723.
Packaging
1, 2 L in Sure/Seal™
18 L in Pure-Pac™ 1
200, 400 L in Pure-Pac™ 1
25 L in ss drum
25 L packaged in closed-head stainless steel drums; 18 L, 56 L, 200 L packaged in Pure-Pac® stainless steel containers; 1 L, 2 L packaged in Sure/Seal™
Legal Information
Sure/Seal is a trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.
Pure-Pac is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.