|
| 612081 Aldrich |
Gadolinium(III) chloride solutionNMR reference standard, 0.1 mg/mL in D2O (99.9 atom % D), DSS 0.1 %, water 1 % |
|
|||||||
| Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. |