Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
652946
Aldrich
4-Bromo-1,3,5-
trimethylpyrazole
puriss., 98%
zoom
1 of 1
Molecular Formula
C
6
H
9
BrN
2
Molecular Weight
189.05
CAS Number
15801-69-1
MDL number
MFCD00052532
Expand/Collapse All
Price and Availability
Descriptions
Packaging
1, 5 g in glass btl
Properties
grade
puriss.
assay
98%
bp
101-102 °C/15 mmHg(lit.)
mp
30-34 °C(lit.)
30-34 °C
Safety
Hazard Codes
Xn
Risk Statements
22
RIDADR
UN 2811 6.1/PG 3
WGK Germany
3
Flash Point(F)
219 °F
Flash Point(C)
103 °C
Related Products
Replaced by
647667
, 4-Bromo-1,3,5-trimethylpyrazole
Related Categories
...
Heterocyclic Building Blocks
>
Pyrazoles
...
Halogenated Heterocycles
>
Pyrazoles
Scroll Up
Scroll Down
Related Information
MSDS
Certificate of Analysis
More Information
Links
Similar Products
Related Products
Related Categories
Page Options
Printer Friendly View
Bulk Quote
Ask A Scientist
Email Page
Last 5 Products Viewed
652946 (Aldrich)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.