Application
A kit of 96 chiral bases for rapid screening of the optimum resolution method for a given racemate.
ChiroSolv Kits are high throughput disposable kits that allow rapid screening to find the optimum resolution method for a given racemate. Each kit offers 96 unique combinations of chiral resolving agents and solvents to screen against. There are 4 kits in the Acid Series and 4 kits in the Base series, allowing for a maximum of 768 combinations. Results can be achieved within 24 hours. Some of the solutes included in this kit are shown below. For a complete list of components, please contact your local Sigma-Aldrich Technical Service office. See Application Guide or More Information links for information on the entire kit range.
Legal Information
ChiroSolv is a registered trademark of ChiroSolve, Inc.