Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
693812
Aldrich
Zirconium-iron alloy
zoom
1 of 1
Synonym
ZrFe
2
Molecular Formula
ZrFe
2
CAS Number
12023-45-9
Expand/Collapse All
Price and Availability
Descriptions
Application
High pressure vessels for moblie storage of hydrogen. New storage systems based on AB2-type intermetallics with good hydrogen absorption parameters. Materials are typical representative of AB2 alloys.
Related Categories
...
Metal and Ceramic Science
>
Alloys
...
New Products for Materials Research and Engineering
>
Alternative Energy
...
Materials for Hydrogen Storage
>
Metal Alloys
Scroll Up
Scroll Down
Related Information
MSDS
Specification Sheet
Certificate of Analysis
More Information
Links
Related Categories
Page Options
Printer Friendly View
Bulk Quote
Ask A Scientist
Email Page
Last 5 Products Viewed
693812 (Aldrich)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.