![]() |
Synonym
2
2
Molecular Formula
2-(H2N)C6H4CO2H
Molecular Weight
137.14
CAS Number
Beilstein Registry Number
471803
MDL number
PubChem Substance ID
EG/EC Number
204-287-5
References
Reference
Huang, Z., et al., The use of the 2-aminobenzoic acid tag for oligosaccharide gel electrophoresis. Carbohydr. Chem. 328, 77-83, (2000)
He, L., et al., Separation of saccharides derivatized with 2-aminobenzoic acid by capillary electrophoresis and their structural consideration by nuclear magnetic resonance. Anal. Biochem. 314, 128-134, (2003)
Neville, D.C., et al., Analysis of fluorescently labeled glycosphingolipid-derived oligosaccharides following ceramide glycanase digestion and anthranilic acid labeling. Anal. Biochem. 331, 275-282, (2004)
Merck
Merck 13,421
Beilstein
Beil. 14,IV,1004
reference
Aldrich MSDS 1, 131:A / Corp MSDS 1 (1), 282:B / FT-IR 2 (2), 2651:B / FT-IR 1 (2), 189:A / FT-NMR 1 (2), 1066:B / IR-Spectra (3), 958:B / IR-Spectra (2), 838:G / NMR-Reference 2 (2), 186:B / RegBook 1 (2), 1783:N / Sax 6, 291 / Sigma FT-IR 1 (2), 291:B / Structure Index 1, 284:D:8
| ||||||||