General description
Serpentine configured heating elements embedded in tough, pliable, braided tape that conforms to almost any shape vessel for even, direct-contact heating without hot spots. All tapes have high temperature lead wires emerging from opposite ends and separable line plugs. A temperature controller is required.
Standard insulated heating tape completely enclosed in a specially compounded, high-strength silicone rubber.
Legal Information
Thermolyne is a registered trademark of Barnstead/Thermolyne Corp.