General description
The FLUKE 177 multimeter is designed to support first level electrical and electronic troubleshooting.
• Single rotary dial selects functions
• LCD display, 3,200 count
• 31-segment analog bar graph
• Touch-Hold* lock function
• Autoranging
• Sleep Mode battery saver
• Diode test
• UL listed
All meters include a pair of test leads, operator's manual, and a 9V battery.
Other Notes
*Touch-Hold feature allows automatic measurements while operator watches probes and circuits. Meter captures measurement, beeps, and locks it in display until operator is ready to view it. Automatically updates with each new measurement.
Legal Information
Fluke is a registered trademark of John Fluke Manufacturing Co., Inc.