General description
New reactor design1 handles volumes from 5 to 15 mL. Borosilicate glass body with #25 Ace-Thred at top, 14/10 outer side joint externally threaded to allow an O-ring cap-seal connection with mating male joints, and a porosity E (4 to 8 µm) filter disc for use as a gas inlet to remove oxygen from the solution or for agitation. Includes an 11 mm I.D. jacketed quartz immersion well, and a quartz Pen-Ray 5.5 W, low pressure, cold cathode, mercury lamp with matching 115 V, 60 Hz power supply.
1.Designed by John H. Penn and Richard D. Orr, West Virginia Univ. Dept. of Chemistry, Morgantown, WV 26506.