General description
Offer a wide variety of membranes that meet almost any filtration need.
Supor® polyethersulfone, for biological and pharmaceutical solutions, fast flow, low extractables, autoclavable
HT Tuffryn® polysulfone, for media and air samples, low-protein-binding, autoclavable
GN Metricel® mixed cellulose esters, for air samples and microbiological analysis, meets US EPA and ASTM specifications, autoclavable
Nylaflo® nylon, for general-purpose applications, HPLC, good chemical resistance, low extractables
PTFE PTFE on PP support, for aggressive organics, HPLC and air/gases, hydrophobic, chemically inert
Legal Information
Supor is a registered trademark of Pall Corp.