|
| Z278068 Aldrich |
Plain-form specific gravity hydrometerfor liquids heavier than water, 1.000-2.000 g/mil specific gravity, accuracy: 0.01 g/mil |
|
|||||||
| Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. |