General description
Derma Shield products are non-greasy, non-toxic, temporary protectants that can be applied to any area of the skin. Within 4 to 5 minutes after proper pplication, Derma Shield becomes an invisible protectant that actually bonds to the skin and helps protect it. The product is applied to skin or hands as a white foam with a mild fragrance.
• Perfect adjunct to wear with gloves, protecting hands in case of failure or leakage
• Allows skin to breathe and perspire normally
• Does not affect sense of touch or natural mobility
• Does not wash off skin once it dries
• Conditions skin with Lanolin, Aloe Vera, and Vitamin E
Packaging
Supplied in a 12 oz aerosol can.
Legal Information
DERMA SHIELD is a registered trademark of Benchmark Commercial, Inc.