General description
• Reduce the risk of laboratory hazards and liability
• UL listed for Class I, Group C hazardous atmospheres
• CE compliant to meet IEC1010 standards
A sturdy, high performance unit that is as dependable as it is safe. Aluminum top plate surface allows for maximum heat transfer and temperature uniformity. Automatic safety switch cuts power if temperature exceeds 246 °C. Temp. range: 38 to 220 °C. Stirring speed: 50 to 1,200 rpm. Load capacity: 25 lb. Includes a built-in ring stand holder. Unit must be hardwired; plug not included.
Legal Information
Thermolyne is a registered trademark of Barnstead/Thermolyne Corp.