|
| Z306800 Aldrich |
Volac® burettevolume 1 mL, accuracy: 0.01 mL, subdivision, 0.01 mL, class A with certificate |
|
|||||||
| Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. |