General description
Blows warm air up tubular supports for rapid drying of laboratory glassware. 56 white plastic-coated lateral drying tubes (24, 16 mm diam. 32 tubes, 11 mm diam.) on rotating central column. Electric heater and fan blows filtered hot or cold air up column and out of tube. Maximum temperature in glass vessel on dryer: approx 50 °C.
Heater top has raised lip to act as condensation/drip tray with drain pipe at rear. Front panel has power and heating indicator lamps, power switch, toggle switch for hot or cold air, and timer.
Legal Information
Gerhardt is a registered trademark of Gerhardt.