General description
0-99 min timer - set/display analog
Interval sieving selectable - Set/display digital
The electromagnetic drives in these analytical sieve shakers have high load endurance, are quiet running, give the sieve 3-dimensional motion, moving the sieving material evenly over the entire sieve surface for short sieving times with precise separation of fractions. For 100, 150, 200 and 203mm diameter sieves. Will collect up to 9 fractions using 50mm high sieves or 17 fractions using 25mm high sieves. Suitable for wet of dry sieving. Supplied as shaker only.
• 0-3mm Vibration amplitude - set/display analog
• Vibration frequency 3000min-1
• Max. sieve stack weight 4kg
• Max. sample weight 3kg