General description
For all types of melting point and boiling point determinations. Units have digital display with simple pushbutton controls, oil-free metal heating blocks, and compact footprints.
• Accuracy: up to 100 °C (±0.3 °C); up to 250 °C (±0.5 °C); up to 400 °C (±0.8 °C)
• 9 heating rates: 0.1, 0.2, 0.5, 1.0, 2.0, 3.0, 5.0, 10.0 and 20.0 °C/min
• Heating: 50 to 350 °C in 7 min; Cooling: 350 to 50 °C in 10 min
• RS-232 interface for data logging according to GLP
• Calibration with WHO reference material possible
• In accordance with DAB10, Ph.Eur. II, Ph.Helv.II regulations
For visual determination of three melting points and two boiling points.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG.