General description
Büchi recirculating chillers offer the environmentally friendly alternative to tap water cooling. In addition, as the cooling medium temperature is -10°C, less solvent is lost during drying or evaporating operations. Two versions, with 800 and 1,400 W cooling power, are available to accommodate either one system or two systems simultaneously. The coolant temperature can be selected according to the specifics of the application. Coolant temperature range: -10 to 40°C.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG.