General description
Self contained refrigeration baths for precise temperature control of external objects in a very wide temperature range. They are also equipped with a bath opening for direct temperature control of immersed samples. The powerful, CFC-free compressor circuit offers a high amount of heat removal capacity, even if working around ambient temperature. Available with only 195 mm width as space saver (K10 types).
C Class are compact analogue models.
DC Class are compact digital models.
Legal Information
Thermo Haake is a registered trademark of Thermo Electron Corp.