General description
These respirators feature the patented Multi-Flex System: a combination of soft, pliable rubber and plastic components for unmatched fit and all-day comfort. Supplied without cartridges. Order Advantage cartridges separately. NIOSH/MSHA approved.
• Economical and low-maintenance
• Superior fit and comfort
• AnthroCurve sealing surface provides superior fit and comfort
• Multi-Flec System AnthroCurve sealing surface equalizes pressure on the face seal area
• Black thermoplastic rubber (TPR) sealing surface with plastic yoke
Legal Information
Advantage is a registered trademark of Mine Safety Appliances Co.