General description
All-in-one meter and built-in rotary vane sensor simultaneously displays air velocity and temperature on large dual LCD display. Up to 2-hour air velocity averaging for measurements. Includes 9V battery and carrying case.
• 4-Digit display of air velocity, accuracy: ±3%
Air velocity, resolution
80 to 5900 ft/min, 1ft/min
0.4 to 30 m/s, 0.01m/s
0.9 to 68 mph, 0.1mph
0.8 to 58 knots, 0.1 knots
1.4 to 108 km/hr, 0.1km/hr
• °C/°F Temperature measurement via built-in thermistor
• Resolution at temperature -10 - 50 °C = 0.1 °C
• Resolution at temperature 14 - 122 °F = 0.1 °F
• Data Hold and Min/Max Record/Recall functions
• Low battery indication and Auto-Power Off
• Internal memory recalls custom configuration