General description
For use with Model RET basic stirring hot plate or any unit with a plate diameter of 135 mm. A pump impeller, driven by the magnetic field of the stirrer, delivers liquid at up to 2 L/min. (reference liquid: H2O). The recirculated liquid is heating up to 180 °C by the hot plate. Heating precision is based on the controls of the stirring hot plate. Recommended use with Model RET basic stirring hot plate, IKATRON electronic thermometer, and PT 1000 short temperature probe.
Legal Information
IKATRON is a registered trademark of IKA Works, Inc.