General description
• Unique vented closure relieves solvent vapor pressure inside wash bottle and eliminates dripping
• Safety information printed on the bottle, including chemical color codes, chemical name and formula, hazard codes, NFPA codes, target organs, and effects and route of entry information
• Color-coded closure
• Easy-to-fill wide neck
• Steady, strong dispensing stream inherent to Unitary bottle design
Legal Information
Unitary is a trademark of Nalge Nunc International.
Nalgene is a registered trademark of Nalge Nunc International.