General description
For simple distillation of solvents and general use. Tap-water-cooled. Inlet feed tube for continuous feed of volumes exceeding flask capacity.
Yamato rotary evaporators offer high performance, reliability, and exceptional quality at an affordable price. They also carry a 24 month warranty (US only). Ideal for use in chemical, pharmaceutical, and R&D laboratories in university and industrial settings.
Yamato Model RE-200 with 3.7 L BM-100 water bath
• Reliable performance at an economical price
• Analog control of bath temperature and rotation
• Optional support arm/jack for easy glassware adjustment
• Power source: 115V AC, 7A evaporator; 6A bath
FEATURES:
• Evaporator flask range: 100 mL to 2 L (standard 1 L)
• Sparkless, high-torque induction motor; speed range: 20 to 180 rpm
• Patented, adjustable 24/40 steam duct
• Water bath temp. range: ambient to 95 °C
• Fluororesin vacuum seal
• Thermal fuse shutoff if bath level falls too low