General description
These coil condensers are designed with an open bottom and can be positioned at any location in the flask just by a turn of the PTFE adapter lock nut. Condenser coils provide increased surface area compared to standard cold-finger condensers. Condensers have quick-disconnect fittings to allow for easy removal of cooling hoses. For use with PTFE adapter Cat. No. Z415243. Supplied complete with (2) quick-disconnect fittings and Viton® O-rings.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.
System 45 is a registered trademark of NDS Technologies, Inc.