General description
Cylindrical style vessel has a 2 mm PTFE valve at the top, with a 14/20 female joint side port. The 32 mm threaded bottom valve assembly incorporates System 45® technology. The coarse fritted disc is easily removed from the PTFE adapter for cleaning and/or replacement. PTFE bottom valve requires 1/4-turn to open or close. Valve stems have a 1/4-28 internal thread in the stem which allows a variety of adapters to be screwed into place. For male 1/4-28 and male Luer adapters, see Cat. No. Z417416 and Z417424.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.
System 45 is a registered trademark of NDS Technologies, Inc.