General description
• Automatic microprocessor controlled temperature ramping for improved reproducibility
• Offers precise melting points up to 400 °C on one to three samples
• Heats from ambient to 250 °C in only 5min
• Cools from 400 to 100 °C in only 3min with built-in fan
• Occupies only 4 × 5 in. of bench space
• Soft rubber eye cushion and ideal height provide comfortable viewing
• Shipped with 100 melting point capillary tubes
Simple Operation
1. Reminder instructions printed right on the front of the unit
2. Apparatus prompts user to select a start temperature, heating rate, and end temperature
3. Unit heats to, and maintains, selected start temperature
4. Audible and visual alarms indicate when oven temperature is stable
5. User inserts capillaries, ramping is initiated by pressing the "RAMP" key
6. Onset and end of melting range is observed via built-in 6X lens and viewing lamp
7. Store melting range values by depressing the "STORE" key
8. High speed fan cools oven back to selected starting temperature to begin next test
Specifications:
Temp. accuracy: ±0.5 °C from 20 to 400 °C
Temp. resolution: 0.1 °C
Ramping rate: adjustable from 0.1 °C/min to 10 °C/min in 0.1 °C increments
Reproducibility: 0.2 °C
Heat-up time: 20 to 250 °C in 5 min
Cool-down time: 400 to 100 °C in 3 min
Use capillary tubes, Z286737, only.
Legal Information
MEL-TEMP is a registered trademark of Barnstead/Thermolyne Corp.