General description
Provides safe, dry heating for clinical and general chemistry applications. Interchangeable heating blocks accommodate test tubes, microcentrifuge tubes, and cuvettes. Order blocks separately.
• Control panel displays "actual" and "set" temperatures
• Non-volatile memory retains temperature settings
• Aluminum and SS housing will not melt or rust
• Massive aluminum heating plate assures even heating
• Platinum RTD probe fits into heating blocks next to samples for precise temperature control
Legal Information
Digi-Block is a registered trademark of Laboratory Devices, Inc.