| Z422193 Aldrich |
Aldrich® five-neck round-bottom flaskcapacity 22,000 mL, center Joint: ST/NS 45/50, side Joints: SJ 65/40 (2) |
|
|||||||||
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. |