General description
"Simultaneously displays air velocity and temperature"
• Large dual display is easy to read
• Fold up protective housing extends to 9 in. for better reach
• Select readouts in ft/min, mph, m/s, knots, and Beaufort force
• Water and shock resistant
• Complete with 3V lithium battery and lanyard
• Air velocity accuracy ± 2%
Air velocity, resolution
60 to 7830 ft/min, 1 ft/min
1.1 to 143 m/s, 0.1 m/s
0.8 to 142 km/h, 0.1 km/h
0.5 to 89 mph, 0.1 mph
0.4 to 77.2 Knots, 0.14 Knots
1 to 17 Beaufort force, 1 Beaufort force
Resolution at temperature -18 - 50 °C = 0.1 °C
Resolution at temperature 0 - 122 °F = 0.1 °F