General description
An autoranging multimeter with advanced functions.
• Measures AC/DC voltage and current, resistance, capacitance, and temperature
• Digital display with 42 segment analog bar graph
• Features include: Range Hold, automatic polarity, and over-range indication
• Functions include: audible continuity, diode, transistor
• RS-232 Interface for data acquisition, display, and report generation
• Complete with test leads, datalogging software and cable, K-type temperature probe, 9 V battery, and rubber holster
DC voltage: 400 mV to 1,000 V, 100 μV, ±(0.5% + 3d)
AC voltage: 400 mV to 750 V, 100 μV, ±(1.0% + 5d)
DC current: 4 mA to 20 A, 100 nA, ±(0.8% + 3d)
AC current: 4 mA to 20 A, 100 nA, ±(1.0% + 5d)
Resistance: 400 Ω to 40 MΩ, 0.1Ω, ±(0.5% + 3d)
Capacitance: 4 nF to 400 nF, 1 pF, ±(3.0% + 5d)
Temperature: 0 to 1,000 °C, 1°, ±(3% + 3d)
Dimensions: 7.4 L × 3.2 W × 1.4 in. D (187 × 81 × 34 mm)